Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010811.2 | NP_034941.2 | 5 - 6 | 1997 | 62 | |
GenBank mRNA | AF074925.1 | - | 3 - 4 | 1247 | 62 | |
AK150100.1 | - | 5 - 6 | 1778 | 62 | ||
AK170298.1 | - | 5 - 6 | 1780 | 62 | ||
AK199361.1 | - | 1 - 2 | 47 | 62 | ||
BC050829.1 | - | 5 - 6 | 1762 | 62 | ||
BC110480.1 | - | 3 - 4 | 1395 | 40048115 | 62 | |
U02304.1 | - | 4 - 5 | 1759 | 62 | ||
X75885.1 | - | 3 - 4 | 1401 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |