Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284392.1 | NP_001271321.1 | 6 - 7 | 725 | 63 | |
NM_001284394.1 | NP_001271323.1 | 7 - 8 | 1022 | 63 | ||
NM_001284398.1 | NP_001271327.1 | 6 - 7 | 709 | 63 | ||
NM_011129.2 | NP_035259.1 | 7 - 8 | 1022 | 63 | ||
GenBank mRNA | AB078010.1 | - | 6 - 7 | 620 | 63 | |
AK133933.1 | - | 6 - 7 | 677 | 63 | ||
AK135143.1 | - | 7 - 8 | 1023 | 63 | ||
BC055101.1 | - | 7 - 8 | 1022 | 6591016 | 63 | |
X61452.1 | - | 7 - 8 | 952 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |