Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pold1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011131.3 | NP_035261.3 | 10 - 11 | 1301 | 71 | |
GenBank mRNA | AK088310.1 | - | 10 - 11 | 1292 | 71 | |
AK146837.1 | - | 10 - 11 | 1298 | 72 | ||
AK166592.1 | - | 10 - 11 | 1495 | 71 | ||
AK167569.1 | - | 10 - 11 | 1284 | 71 | ||
AK168967.1 | - | 10 - 11 | 1264 | 71 | ||
AK169040.1 | - | 10 - 11 | 1297 | 71 | ||
BC009128.1 | - | 10 - 11 | 1302 | 3257174 | 71 | |
BC052670.1 | - | 9 - 10 | 1268 | 30058314 | 71 | |
Z21848.1 | - | 10 - 11 | 1270 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |