Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011201.3 | NP_035331.3 | 3 - 4 | 426 | 61 | |
GenBank mRNA | AK149979.1 | - | 3 - 4 | 382 | 61 | |
AK150322.1 | - | 3 - 4 | 382 | 61 | ||
AK151371.1 | - | 3 - 4 | 339 | 61 | ||
AK153088.1 | - | 3 - 4 | 365 | 61 | ||
AK156281.1 | - | 3 - 4 | 387 | 61 | ||
AK157431.1 | - | 3 - 4 | 427 | 61 | ||
AK165567.1 | - | 3 - 4 | 391 | 61 | ||
AK165949.1 | - | 4 - 5 | 506 | 61 | ||
AK166020.1 | - | 3 - 4 | 382 | 61 | ||
AK168772.1 | - | 3 - 4 | 381 | 61 | ||
AK170620.1 | - | 3 - 4 | 382 | 61 | ||
AK171380.1 | - | 3 - 4 | 382 | 61 | ||
AK172206.1 | - | 4 - 5 | 499 | 61 | ||
AK185938.1 | - | 2 - 3 | 133 | 61 | ||
BC010191.1 | - | 3 - 4 | 344 | 3156854 | 61 | |
M97590.1 | - | 3 - 4 | 342 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |