Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166381.1 | NP_001159853.1 | 6 - 7 | 616 | 109 | |
NM_001166382.1 | NP_001159854.1 | 6 - 7 | 616 | 109 | ||
NM_001166383.1 | NP_001159855.1 | 6 - 7 | 616 | 109 | ||
NM_011236.2 | NP_035366.2 | 6 - 7 | 616 | 109 | ||
GenBank mRNA | AK080702.1 | - | 6 - 7 | 589 | 109 | |
BC010673.1 | - | 5 - 6 | 488 | 109 | ||
BC019123.1 | - | 6 - 7 | 596 | 5003696 | 109 | |
U06837.1 | - | 6 - 7 | 538 | 109 | ||
U12135.1 | - | 6 - 7 | 585 | 109 | ||
Z32767.1 | - | 6 - 7 | 610 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |