Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ski | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011385.2 | NP_035515.2 | 2 - 3 | 1113 | 54 | |
GenBank mRNA | AF435852.1 | - | 2 - 3 | 1092 | 54 | |
AK019148.2 | - | 2 - 3 | 170 | 54 | ||
BC068305.1 | - | 3 - 4 | 957 | 30093626 | 54 | |
CA451127.1 | - | 2 - 3 | 437 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |