Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P116372 |
BC025931.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P116372 |
NM_146064.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
505944 |
AK080166.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
505944 |
M35525.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
505944 |
M35526.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
505944 |
NM_010406.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
254119 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
770907 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
891884 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
794195 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
666804 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
219838 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
154629 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
96548 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P101052 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P134547 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P116372 |
BC025931.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P116372 |
NM_146064.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P196997 |
AF078751.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P196997 |
BC025931.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P196997 |
NM_146064.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30496303 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30496288 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30496295 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30496289 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30496296 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30496291 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30496298 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30496305 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30496293 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30496301 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30496294 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P134547 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P101052 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P196997 |
AF078751.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P196997 |
BC025931.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P196997 |
NM_146064.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P116372 |
BC025931.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P116372 |
NM_146064.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P137825 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P101052 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P134547 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12169480 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12169495 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12169490 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12169481 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12169491 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496293 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496298 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496336 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496390 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496349 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496326 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496358 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496330 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496258 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496263 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496344 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496360 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496368 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496327 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496291 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496288 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496238 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496265 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496320 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496376 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496356 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496311 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496388 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496340 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496319 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496374 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496262 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496354 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496270 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496367 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496235 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496380 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496294 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496314 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496308 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496271 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496372 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496255 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496246 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496241 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496267 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496248 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496333 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496252 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496232 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496243 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496296 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496345 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496346 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496260 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496378 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496375 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496335 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496338 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496256 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496317 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496364 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496289 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496250 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496306 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496295 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496254 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30496347 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30496291 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12169478 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30496328 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12169494 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30496374 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11983206 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12457079 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30496372 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12392981 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12019816 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30496304 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30496317 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30496302 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30496326 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30496327 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12169493 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30496309 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30496379 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12197233 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30496378 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30496330 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30496320 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12322167 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30496311 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30496301 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30496288 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30496306 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30496315 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12237434 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30496289 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30496331 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12265602 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12169498 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30496376 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30496370 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30496322 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30496303 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30496294 |
- |
- |