Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011550.3 | NP_035680.3 | 1 - 2 | 244 | 115 | |
GenBank mRNA | AF213672.1 | - | 1 - 2 | 208 | 115 | |
BC031380.1 | - | 1 - 2 | 224 | 5252695 | 115 | |
BC038004.1 | - | 1 - 2 | 224 | 115 | ||
U43548.1 | - | 1 - 2 | 204 | 115 | ||
U43607.1 | - | 1 - 2 | 215 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |