Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011580.3 | NP_035710.2 | 15 - 16 | 2596 | 59 | |
GenBank mRNA | AK080686.1 | - | 15 - 16 | 2631 | 59 | |
AK145202.1 | - | 15 - 16 | 2626 | 59 | ||
AK163092.1 | - | 15 - 16 | 2623 | 59 | ||
BC042422.1 | - | 15 - 16 | 2598 | 4018176 | 59 | |
BC050917.1 | - | 15 - 16 | 2596 | 6405710 | 59 | |
M87276.1 | - | 15 - 16 | 2622 | 59 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AK089315.1 | - | Chr. - - [-] on Build | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |