Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163664.1 | NP_001157136.1 | 11 - 12 | 297 | 88 | |
NM_001163665.1 | NP_001157137.1 | 11 - 12 | 297 | 88 | ||
NM_001163666.1 | NP_001157138.1 | 9 - 10 | 270 | 88 | ||
NM_001163667.1 | NP_001157139.1 | 10 - 11 | 285 | 88 | ||
NM_001163668.1 | NP_001157140.1 | 9 - 10 | 246 | 88 | ||
NM_001163669.1 | NP_001157141.1 | 10 - 11 | 258 | 88 | ||
NM_001163670.1 | NP_001157142.1 | 7 - 8 | 213 | 88 | ||
NM_011620.3 | NP_035750.2 | 8 - 9 | 225 | 88 | ||
GenBank mRNA | AA028290.1 | - | 10 - 11 | 238 | 88 | |
AA754928.1 | - | 8 - 9 | 201 | 87 | ||
AA771596.1 | - | 9 - 10 | 223 | 87 | ||
AI173874.1 | - | 9 - 10 | 262 | 88 | ||
AI574395.1 | - | 11 - 12 | 291 | 88 | ||
AI746973.1 | - | 9 - 10 | 265 | 88 | ||
AK003147.1 | - | 7 - 8 | 180 | 88 | ||
BC003747.1 | - | 11 - 12 | 293 | 3594256 | 88 | |
BY766323.1 | - | 10 - 11 | 275 | 88 | ||
DV049834.1 | - | 8 - 9 | 272 | 88 | ||
L48988.1 | - | 5 - 6 | 116 | 88 | ||
L48989.1 | - | 5 - 6 | 116 | 88 | ||
L48990.1 | - | 8 - 9 | 161 | 88 | ||
L48991.1 | - | 5 - 6 | 143 | 88 | ||
L48992.1 | - | 7 - 8 | 149 | 88 | ||
L48993.1 | - | 7 - 8 | 149 | 88 | ||
L49018.1 | - | 5 - 6 | 143 | 88 | ||
L49466.1 | - | 10 - 11 | 285 | 88 | ||
L49467.1 | - | 11 - 12 | 297 | 88 | ||
L49468.1 | - | 9 - 10 | 270 | 88 | ||
L49469.1 | - | 9 - 10 | 270 | 88 | ||
L49470.1 | - | 10 - 11 | 258 | 88 | ||
L49471.1 | - | 9 - 10 | 267 | 88 | ||
L49472.1 | - | 9 - 10 | 246 | 88 | ||
U77779.1 | - | 8 - 9 | 218 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |