Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011875.4 | NP_036005.1 | 3 - 4 | 293 | 118 | |
GenBank mRNA | AF107838.1 | - | 3 - 4 | 254 | 118 | |
AK051078.1 | - | 3 - 4 | 284 | 118 | ||
AK075936.1 | - | 3 - 4 | 294 | 118 | ||
AK081160.1 | - | 3 - 4 | 291 | 118 | ||
AK145654.1 | - | 3 - 4 | 256 | 118 | ||
AK163017.1 | - | 3 - 4 | 288 | 118 | ||
AK167169.1 | - | 3 - 4 | 278 | 118 | ||
AK171504.1 | - | 3 - 4 | 274 | 118 | ||
AK206994.1 | - | 3 - 4 | 169 | 118 | ||
BC029989.1 | - | 3 - 4 | 233 | 118 | ||
BC139184.1 | - | 3 - 4 | 271 | 8862206 | 118 | |
BC139187.1 | - | 3 - 4 | 271 | 8862209 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |