Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poli | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011972.2 | NP_036102.2 | 1 - 2 | 131 | 79 | |
GenBank mRNA | AF151691.1 | - | 1 - 2 | 131 | 79 | |
AK145244.1 | - | 1 - 2 | 76 | 79 | ||
HQ698885.1 | - | 1 - 2 | 264 | 79 | ||
HQ698886.1 | - | 1 - 2 | 264 | 79 | ||
HQ698887.1 | - | 1 - 2 | 264 | 79 | ||
HQ698888.1 | - | 1 - 2 | 264 | 79 | ||
HQ698889.1 | - | 1 - 2 | 264 | 79 | ||
HQ698890.1 | - | 1 - 2 | 264 | 79 | ||
HQ698891.1 | - | 1 - 2 | 264 | 79 | ||
HQ698892.1 | - | 1 - 2 | 264 | 79 | ||
HQ698893.1 | - | 1 - 2 | 264 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus 1700 Inventoried Probe spans exons |