Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011986.4 | NP_036116.3 | 4 - 5 | 1815 | 82 | |
GenBank mRNA | AK002938.1 | - | 2 - 3 | 420 | 82 | |
AK122327.1 | - | 2 - 3 | 1160 | 82 | ||
AK149932.1 | - | 2 - 3 | 420 | 82 | ||
AK170502.1 | - | 5 - 6 | 1530 | 82 | ||
BC017126.1 | - | 4 - 5 | 1746 | 4485839 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |