Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163588.1 | NP_001157060.1 | 3 - 4 | 747 | 99 | |
NM_001163589.1 | NP_001157061.1 | 3 - 4 | 747 | 99 | ||
NM_013639.2 | NP_038667.2 | 3 - 4 | 747 | 99 | ||
GenBank mRNA | AK149021.1 | - | 3 - 4 | 749 | 99 | |
BC046291.1 | - | 3 - 4 | 740 | 6475506 | 99 | |
X15475.1 | - | 3 - 4 | 509 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |