Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013651.4 | NP_038679.3 | 5 - 6 | 464 | 79 | |
GenBank mRNA | BC052697.1 | - | 5 - 6 | 433 | 5715282 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |