Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Podxl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013723.3 | NP_038751.2 | 2 - 3 | 936 | 104 | |
GenBank mRNA | AB028048.1 | - | 2 - 3 | 817 | 104 | |
AF290209.1 | - | 2 - 3 | 835 | 104 | ||
BC052442.1 | - | 2 - 3 | 910 | 6406610 | 104 | |
BC054530.1 | - | 2 - 3 | 910 | 6406610 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |