Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013752.3 | NP_038780.3 | 8 - 9 | 1094 | 97 | |
GenBank mRNA | AB016988.1 | - | 8 - 9 | 1016 | 97 | |
AF076687.1 | - | 8 - 9 | 1088 | 97 | ||
AF092840.1 | - | 8 - 9 | 1077 | 97 | ||
AK031933.1 | - | 8 - 9 | 1080 | 97 | ||
AK132302.1 | - | 8 - 9 | 1135 | 97 | ||
AK134805.1 | - | 8 - 9 | 1130 | 97 | ||
AK134960.1 | - | 8 - 9 | 1122 | 97 | ||
AK159790.1 | - | 8 - 9 | 1101 | 97 | ||
AK160614.1 | - | 8 - 9 | 1061 | 97 | ||
BC003719.1 | - | 8 - 9 | 1103 | 97 | ||
BC013190.1 | - | 8 - 9 | 1094 | 97 | ||
BC044773.2 | - | 8 - 9 | 1093 | 5067851 | 97 | |
BC055061.1 | - | 8 - 9 | 1086 | 6528890 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |