Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159284.1 | NP_001152756.1 | 15 - 16 | 2312 | 63 | |
NM_001284427.1 | NP_001271356.1 | 14 - 15 | 2261 | 63 | ||
NM_001284428.1 | NP_001271357.1 | 15 - 16 | 2369 | 63 | ||
NM_001284429.1 | NP_001271358.1 | 15 - 16 | 2314 | 63 | ||
NM_013870.3 | NP_038898.2 | 14 - 15 | 2261 | 63 | ||
GenBank mRNA | AF064236.1 | - | 14 - 15 | 2208 | 63 | |
AJ010305.1 | - | 14 - 15 | 2208 | 63 | ||
AK030932.1 | - | 14 - 15 | 2262 | 63 | ||
AK083947.1 | - | 4 - 5 | 515 | 63 | ||
AK085412.1 | - | 15 - 16 | 2277 | 63 | ||
AK138898.1 | - | 15 - 16 | 2327 | 63 | ||
AK160492.1 | - | 5 - 6 | 716 | 63 | ||
AK170516.1 | - | 14 - 15 | 2221 | 63 | ||
BC010599.1 | - | 14 - 15 | 2202 | 3709078 | 63 | |
BC066192.1 | - | 15 - 16 | 2187 | 30433616 | 63 | |
BC069836.1 | - | 15 - 16 | 2187 | 30456568 | 63 | |
BC069839.1 | - | 15 - 16 | 2187 | 30431675 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |