Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013874.2 | NP_038902.1 | 8 - 9 | 860 | 78 | |
GenBank mRNA | BC052348.1 | - | 8 - 9 | 889 | 6587002 | 78 |
U48238.1 | - | 8 - 9 | 859 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |