Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scarb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205082.1 | NP_001192011.1 | 4 - 5 | 835 | 68 | |
NM_001205083.1 | NP_001192012.1 | 4 - 5 | 835 | 68 | ||
NM_016741.2 | NP_058021.1 | 4 - 5 | 835 | 68 | ||
GenBank mRNA | AK010622.1 | - | 4 - 5 | 760 | 68 | |
AK028191.1 | - | 4 - 5 | 816 | 68 | ||
AK033114.1 | - | 4 - 5 | 760 | 68 | ||
AK145588.1 | - | 4 - 5 | 761 | 68 | ||
AK154933.1 | - | 4 - 5 | 827 | 68 | ||
AK158122.1 | - | 4 - 5 | 760 | 68 | ||
AK169906.1 | - | 4 - 5 | 811 | 68 | ||
AK171602.1 | - | 4 - 5 | 815 | 68 | ||
BC004656.1 | - | 4 - 5 | 815 | 3498018 | 68 | |
CT010222.1 | - | 4 - 5 | 623 | 68 | ||
U37799.1 | - | 4 - 5 | 673 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |