Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016985.2 | NP_058681.1 | 8 - 9 | 1032 | 98 | |
GenBank mRNA | AF073997.1 | - | 8 - 9 | 1005 | 98 | |
BC056376.1 | - | 8 - 9 | 1032 | 6849757 | 98 | |
BC057337.1 | - | 8 - 9 | 1026 | 6825916 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |