Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ostf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017375.3 | NP_059071.1 | 4 - 5 | 356 | 76 | |
GenBank mRNA | AK002899.1 | - | 4 - 5 | 335 | 76 | |
AK005525.1 | - | 4 - 5 | 325 | 76 | ||
AK008123.1 | - | 4 - 5 | 304 | 76 | ||
AK008467.1 | - | 4 - 5 | 304 | 76 | ||
AK010074.1 | - | 4 - 5 | 352 | 76 | ||
AK145949.1 | - | 4 - 5 | 307 | 76 | ||
AK149175.1 | - | 76 | ||||
AK150145.1 | - | 4 - 5 | 326 | 76 | ||
AK155028.1 | - | 4 - 5 | 326 | 76 | ||
BC060986.1 | - | 4 - 5 | 235 | 30300544 | 76 | |
U58888.1 | - | 4 - 5 | 304 | 76 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BC020352.1 | - | Chr. - - [-] on Build | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |