Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100018640.mm.1 |
NM_001113486.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018640.mm.1 |
NM_001113486.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018640.mm.1 |
NM_001113487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018640.mm.1 |
NM_001113487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018640.mm.1 |
NM_001113488.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018640.mm.1 |
NM_001113488.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018640.mm.1 |
NM_017380.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018640.mm.1 |
NM_017380.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018640.mm.1 |
TC1100001956.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100018636.mm.1 |
NM_017380.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018636.mm.1 |
NM_017380.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018636.mm.1 |
TC1100001956.mm.1 |
MTA TC |