Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113486.1 | NP_001106958.1 | 10 - 11 | 1661 | 73 | |
NM_001113487.1 | NP_001106959.1 | 9 - 10 | 1841 | 73 | ||
NM_001113488.1 | NP_001106960.1 | 8 - 9 | 1010 | 73 | ||
NM_017380.2 | NP_059076.1 | 8 - 9 | 991 | 73 | ||
GenBank mRNA | AJ250723.1 | - | 8 - 9 | 997 | 73 | |
AK031757.1 | - | 8 - 9 | 886 | 73 | ||
AK122415.2 | - | 9 - 10 | 1635 | 73 | ||
AK141312.1 | - | 10 - 11 | 1602 | 73 | ||
BC046524.1 | - | 10 - 11 | 1661 | 5290621 | 73 | |
BC094455.1 | - | 11 - 12 | 1365 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |