Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St3gal6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018784.2 | NP_061254.1 | 6 - 7 | 831 | 81 | |
GenBank mRNA | AB063326.1 | - | 4 - 5 | 334 | 81 | |
AF119390.1 | - | 6 - 7 | 831 | 81 | ||
AK028442.1 | - | 6 - 7 | 650 | 81 | ||
AK033562.1 | - | 6 - 7 | 718 | 81 | ||
AK041173.1 | - | 6 - 7 | 874 | 81 | ||
AK048950.1 | - | 6 - 7 | 875 | 81 | ||
AK162619.1 | - | 7 - 8 | 811 | 81 | ||
AK197012.1 | - | 2 - 3 | 107 | 81 | ||
BC052338.1 | - | 5 - 6 | 592 | 30061291 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |