Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St3gal6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018784.2 | NP_061254.1 | 9 - 10 | 1252 | 91 | |
GenBank mRNA | AB063326.1 | - | 7 - 8 | 755 | 91 | |
AF119390.1 | - | 9 - 10 | 1252 | 91 | ||
AK028442.1 | - | 9 - 10 | 1072 | 91 | ||
AK033562.1 | - | 9 - 10 | 1139 | 91 | ||
AK048950.1 | - | 9 - 10 | 1296 | 91 | ||
AK162619.1 | - | 10 - 11 | 1232 | 91 | ||
AK214658.1 | - | 1 - 2 | 140 | 91 | ||
AK214980.1 | - | 2 - 3 | 181 | 91 | ||
BC052338.1 | - | 8 - 9 | 1013 | 30061291 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |