Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rev1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019570.3 | NP_062516.2 | 16 - 17 | 2826 | 74 | |
GenBank mRNA | AB057418.1 | - | 15 - 16 | 2615 | 74 | |
AF179302.1 | - | 15 - 16 | 2615 | 74 | ||
AK048838.1 | - | 16 - 17 | 2830 | 74 | ||
BC006721.1 | - | 11 - 12 | 2150 | 74 | ||
BC058093.1 | - | 16 - 17 | 2826 | 6827108 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |