Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252580.1 | NP_001239509.1 | 17 - 18 | 3181 | 63 | |
NM_019729.3 | NP_062703.2 | 17 - 18 | 3148 | 63 | ||
GenBank mRNA | AB045709.1 | - | 16 - 17 | 2922 | 63 | |
AF057146.1 | - | 16 - 17 | 2939 | 63 | ||
AK122195.1 | - | 17 - 18 | 3059 | 63 | ||
AK129003.1 | - | 17 - 18 | 3149 | 63 | ||
AK134058.1 | - | 17 - 18 | 3103 | 63 | ||
AK142881.1 | - | 14 - 15 | 2646 | 63 | ||
AK153015.1 | - | 16 - 17 | 2895 | 63 | ||
AK166540.1 | - | 15 - 16 | 2502 | 63 | ||
AK204074.1 | - | 1 - 2 | 63 | 63 | ||
BC027052.1 | - | 7 - 8 | 1701 | 63 | ||
BC050947.1 | - | 17 - 18 | 3113 | 6825981 | 63 | |
BC061465.1 | - | 17 - 18 | 3113 | 6825981 | 63 | |
BC066126.1 | - | 17 - 18 | 3111 | 30301665 | 63 | |
BC092078.1 | - | 5 - 6 | 941 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |