Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001141931.1 | NP_001135403.1 | 12 - 13 | 1380 | 112 | |
NM_001141932.1 | NP_001135404.1 | 13 - 14 | 1428 | 112 | ||
NM_020296.2 | NP_064692.2 | 12 - 13 | 1573 | 112 | ||
GenBank mRNA | AB026569.1 | - | 12 - 13 | 1296 | 112 | |
AK133054.1 | - | 12 - 13 | 1101 | 112 | ||
AK135280.1 | - | 12 - 13 | 1260 | 112 | ||
AK145897.1 | - | 12 - 13 | 1382 | 112 | ||
AK147109.1 | - | 12 - 13 | 1574 | 112 | ||
AK161102.1 | - | 13 - 14 | 1378 | 112 | ||
AK161416.1 | - | 8 - 9 | 710 | 112 | ||
BC016501.1 | - | 12 - 13 | 1561 | 4527742 | 112 | |
BC056609.1 | - | 10 - 11 | 820 | 112 | ||
BC057866.1 | - | 13 - 14 | 1460 | 5327143 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |