Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146001.1 | NP_001139473.1 | 2 - 3 | 254 | 84 | |
NM_021517.2 | NP_067492.2 | 2 - 3 | 300 | 84 | ||
GenBank mRNA | AF220100.1 | - | 2 - 3 | 239 | 84 | |
AK006269.1 | - | 2 - 3 | 774 | 84 | ||
AK012070.1 | - | 2 - 3 | 290 | 84 | ||
AK014879.1 | - | 2 - 3 | 557 | 84 | ||
AK029752.1 | - | 2 - 3 | 758 | 84 | ||
AK029764.1 | - | 2 - 3 | 768 | 84 | ||
AK165415.1 | - | 2 - 3 | 235 | 84 | ||
AW259368.1 | - | 2 - 3 | 300 | 84 | ||
BC013512.1 | - | 1 - 2 | 223 | 4234949 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |