Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nampt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021524.2 | NP_067499.2 | 1 - 2 | 290 | 79 | |
GenBank mRNA | AF234625.1 | - | 1 - 2 | 150 | 79 | |
AK086415.1 | - | 1 - 2 | 121 | 79 | ||
AK153926.1 | - | 1 - 2 | 121 | 79 | ||
AK166270.1 | - | 1 - 2 | 150 | 79 | ||
AY679720.1 | - | 1 - 2 | 125 | 79 | ||
BC004059.1 | - | 1 - 2 | 98 | 3590087 | 79 | |
BC018358.1 | - | 1 - 2 | 290 | 2609271 | 79 | |
BY068469.1 | - | 1 - 2 | 282 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |