Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nampt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021524.2 | NP_067499.2 | 2 - 3 | 455 | 108 | |
GenBank mRNA | AF234625.1 | - | 2 - 3 | 315 | 108 | |
AK086415.1 | - | 2 - 3 | 286 | 108 | ||
AK153926.1 | - | 2 - 3 | 286 | 108 | ||
AK166270.1 | - | 2 - 3 | 315 | 108 | ||
AY679720.1 | - | 2 - 3 | 290 | 108 | ||
BC004059.1 | - | 2 - 3 | 263 | 3590087 | 108 | |
BC018358.1 | - | 2 - 3 | 455 | 2609271 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |