Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncstn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021607.3 | NP_067620.3 | 3 - 4 | 374 | 106 | |
GenBank mRNA | AF240469.1 | - | 3 - 4 | 324 | 106 | |
AK033314.1 | - | 3 - 4 | 316 | 106 | ||
AK129102.1 | - | 3 - 4 | 374 | 106 | ||
AK170173.1 | - | 3 - 4 | 320 | 106 | ||
AK170244.1 | - | 3 - 4 | 339 | 106 | ||
AK170369.1 | - | 3 - 4 | 363 | 106 | ||
AK172579.1 | - | 3 - 4 | 319 | 106 | ||
BC019998.1 | - | 3 - 4 | 319 | 4458780 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |