Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Twsg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023053.3 | NP_075540.1 | 2 - 3 | 281 | 91 | |
GenBank mRNA | AF292033.1 | - | 220 | 91 | ||
AF295097.1 | - | 1 - 2 | 139 | 91 | ||
AJ297390.1 | - | 2 - 3 | 231 | 91 | ||
AK033504.1 | - | 2 - 3 | 240 | 91 | ||
AK049646.1 | - | 2 - 3 | 275 | 91 | ||
AK168547.1 | - | 2 - 3 | 231 | 91 | ||
AK168633.1 | - | 2 - 3 | 240 | 91 | ||
BC004850.1 | - | 2 - 3 | 236 | 2810960 | 91 | |
CJ165542.1 | - | 2 - 3 | 282 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |