Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Palm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161747.1 | NP_001155219.1 | 3 - 4 | 306 | 94 | |
NM_023128.4 | NP_075617.3 | 3 - 4 | 306 | 94 | ||
GenBank mRNA | AK009200.1 | - | 2 - 3 | 147 | 94 | |
AK037044.1 | - | 3 - 4 | 163 | 94 | ||
AK082163.1 | - | 3 - 4 | 308 | 94 | ||
AK129107.1 | - | 1 - 2 | 45 | 94 | ||
AK133733.1 | - | 3 - 4 | 212 | 94 | ||
BC015297.1 | - | 3 - 4 | 290 | 4223845 | 94 | |
Y14771.1 | - | 3 - 4 | 269 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |