Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scyl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023912.2 | NP_076401.1 | 9 - 10 | 1461 | 78 | |
GenBank mRNA | AF276514.1 | - | 9 - 10 | 1461 | 78 | |
AK019240.1 | - | 9 - 10 | 1454 | 78 | ||
AK134356.1 | - | 9 - 10 | 1460 | 78 | ||
AK144134.1 | - | 9 - 10 | 1447 | 78 | ||
AK145869.1 | - | 9 - 10 | 1481 | 78 | ||
AK161813.1 | - | 9 - 10 | 1460 | 78 | ||
BC034519.1 | - | 9 - 10 | 1447 | 3672524 | 78 | |
HM856325.1 | - | 9 - 10 | 1461 | 78 | ||
HM856326.1 | - | 9 - 10 | 1461 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |