Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024281.2 | NP_077243.2 | 1 - 2 | 267 | 97 | |
NM_133626.2 | NP_598329.1 | 1 - 2 | 267 | 97 | ||
GenBank mRNA | AF273683.1 | - | 190 | 97 | ||
AF273684.1 | - | 1 - 2 | 190 | 97 | ||
AF273685.1 | - | 1 - 2 | 190 | 97 | ||
AF273686.1 | - | 1 - 2 | 190 | 97 | ||
AF273687.1 | - | 1 - 2 | 190 | 97 | ||
AF273688.1 | - | 1 - 2 | 190 | 97 | ||
AF273689.1 | - | 1 - 2 | 190 | 97 | ||
AF273690.1 | - | 1 - 2 | 190 | 97 | ||
AF273691.1 | - | 1 - 2 | 190 | 97 | ||
AF273692.1 | - | 1 - 2 | 190 | 97 | ||
AF273693.1 | - | 1 - 2 | 190 | 97 | ||
AK039114.1 | - | 1 - 2 | 267 | 97 | ||
BC002105.1 | - | 1 - 2 | 177 | 97 | ||
BC031452.1 | - | 1 - 2 | 256 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |