Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24586142 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24586143 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24586140 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P604526 |
BC020315.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P604526 |
BC043921.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P604526 |
AK050637.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P604526 |
AK141554.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P604526 |
BC061064.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P604526 |
BC020315.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P604526 |
BC043921.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P604526 |
AK050637.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P604526 |
AK141554.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P604526 |
BC061064.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
729176 |
AK020480.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
179715 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485442 |
AK048568.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485442 |
BC007166.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485442 |
BC011099.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485442 |
BC013557.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485442 |
BC089158.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485442 |
D28818.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485442 |
NM_013512.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
376583 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
303208 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
141494 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P129736 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P167745 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P604526 |
BC020315.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P604526 |
BC043921.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P604526 |
AK050637.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P604526 |
AK141554.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P604526 |
BC061064.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P604526 |
BC020315.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P604526 |
BC043921.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P604526 |
AK050637.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P604526 |
AK141554.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P604526 |
BC061064.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24586133 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24586132 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24586156 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24586162 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24586127 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24586135 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24586142 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24586128 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24586136 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24586153 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24586130 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24586148 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24586160 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24586143 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24586154 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24586125 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24586145 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24586161 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24586139 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24586146 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24586150 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24586158 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P167745 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P604526 |
BC020315.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P604526 |
BC043921.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P604526 |
AK050637.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P604526 |
AK141554.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P604526 |
BC061064.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P604526 |
BC020315.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P604526 |
BC043921.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P604526 |
AK050637.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P604526 |
AK141554.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P604526 |
BC061064.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P104565 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P167745 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05817882 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05817884 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05817888 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24586140 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24586142 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24586143 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586176 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586169 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586153 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586171 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586093 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586162 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586104 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586161 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586096 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586128 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586119 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586101 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586120 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586121 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586094 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586108 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586106 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586102 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586090 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586132 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586109 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586092 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586130 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586133 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586112 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586127 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586105 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586115 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586117 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586139 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586089 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586099 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586150 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586145 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586142 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586125 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586151 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586173 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586143 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586124 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586164 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24586136 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586117 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586158 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05079298 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586130 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586106 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586096 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05120495 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05874642 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05656469 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586139 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05260717 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05804597 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586156 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05260719 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586162 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05817882 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586153 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586122 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586135 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05740390 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586159 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586093 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586124 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586112 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586155 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586119 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586103 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586120 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586125 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05656466 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586145 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586123 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586099 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586150 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586161 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05274011 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05817884 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586105 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586128 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05837959 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586102 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586136 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05656472 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24586101 |
- |
- |