Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026744.3 | NP_081020.1 | 1 - 2 | 99 | 82 | |
GenBank mRNA | AK003549.1 | - | 1 - 2 | 100 | 82 | |
BC022162.1 | - | 1 - 2 | 124 | 5148541 | 82 | |
BC065995.1 | - | 1322 | 5688315 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons |