Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027949.1 | NP_082225.1 | 2 - 3 | 624 | 95 | |
GenBank mRNA | AK005673.1 | - | 2 - 3 | 661 | 95 | |
AK005852.1 | - | 2 - 3 | 624 | 95 | ||
AK148191.1 | - | 2 - 3 | 707 | 95 | ||
AK155352.1 | - | 2 - 3 | 425 | 95 | ||
BC049113.1 | - | 2 - 3 | 721 | 95 | ||
BC057174.1 | - | 2 - 3 | 471 | 5004450 | 95 | |
BC132517.1 | - | 2 - 3 | 414 | 40130794 | 95 | |
BC138571.1 | - | 2 - 3 | 414 | 8861591 | 95 | |
BC145337.2 | - | 2 - 3 | 414 | 9053893 | 95 | |
BC145338.1 | - | 2 - 3 | 414 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |