Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Robo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF536772.1 | - | 16 - 17 | 2861 | 76 | |
AK004723.2 | - | 16 - 17 | 2837 | 76 | ||
AK030973.1 | - | 1628 | 76 | |||
AK087355.1 | - | 15 - 16 | 3013 | 76 | ||
AK172104.1 | - | 15 - 16 | 3099 | 76 | ||
BC020129.1 | - | 8 - 9 | 1381 | 76 | ||
BC071193.1 | - | 15 - 16 | 2551 | 5060473 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |