Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcolce2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029620.2 | NP_083896.1 | 1 - 2 | 247 | 82 | |
GenBank mRNA | AF352788.1 | - | 1 - 2 | 143 | 82 | |
AK010249.1 | - | 1 - 2 | 248 | 82 | ||
AK132395.1 | - | 137 | 82 | |||
BC051174.1 | - | 1 - 2 | 120 | 6310294 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |