Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122829.1 | NP_001116301.1 | 17 - 18 | 2754 | 108 | |
NM_030680.2 | NP_109605.2 | 17 - 18 | 2721 | 108 | ||
GenBank mRNA | AF182947.1 | - | 18 - 19 | 108 | ||
AF322655.1 | - | 17 - 18 | 108 | |||
AK148196.1 | - | 18 - 19 | 108 | |||
BC030916.1 | - | 5 - 6 | 732 | 108 | ||
BC052149.1 | - | 17 - 18 | 2659 | 6332288 | 108 | |
BC056442.1 | - | 17 - 18 | 2754 | 5702493 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |