Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1800011805.mm.1 |
NM_030683.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011805.mm.1 |
NM_030683.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011805.mm.1 |
NM_207651.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011805.mm.1 |
NM_207651.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011805.mm.1 |
TC1800001638.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800011804.mm.1 |
NM_030683.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011804.mm.1 |
NM_030683.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011804.mm.1 |
NM_207651.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011804.mm.1 |
NM_207651.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011804.mm.1 |
TC1800001638.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800011806.mm.1 |
NM_030683.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011806.mm.1 |
NM_030683.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011806.mm.1 |
NM_207651.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011806.mm.1 |
NM_207651.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011806.mm.1 |
TC1800001638.mm.1 |
MTA TC |