Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tktl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031379.2 | NP_113556.2 | 11 - 12 | 1520 | 96 | |
GenBank mRNA | AF285571.1 | - | 11 - 12 | 1495 | 96 | |
AJ627042.1 | - | 4 - 5 | 410 | 96 | ||
AK029546.1 | - | 11 - 12 | 1521 | 96 | ||
AK139841.1 | - | 11 - 12 | 1520 | 96 | ||
BC113768.1 | - | 11 - 12 | 1495 | 40058456 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |