Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mvp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080638.3 | NP_542369.2 | 3 - 4 | 420 | 80 | |
GenBank mRNA | AF210456.1 | - | 3 - 4 | 422 | 80 | |
AK088046.1 | - | 3 - 4 | 405 | 80 | ||
AK152534.1 | - | 3 - 4 | 395 | 80 | ||
AK167899.1 | - | 3 - 4 | 396 | 80 | ||
AK168100.1 | - | 3 - 4 | 396 | 80 | ||
BC006709.1 | - | 3 - 4 | 385 | 3584439 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |