Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290651.1 | NP_001277580.1 | 12 - 13 | 1673 | 75 | |
NM_001290652.1 | NP_001277581.1 | 12 - 13 | 1673 | 75 | ||
GenBank mRNA | AF285577.1 | - | 14 - 15 | 1499 | 75 | |
AK016533.1 | - | 15 - 16 | 1545 | 75 | ||
AK031490.1 | - | 12 - 13 | 1652 | 75 | ||
AK040521.1 | - | 12 - 13 | 1675 | 75 | ||
BC056382.1 | - | 12 - 13 | 1360 | 6853936 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |