Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spats2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139140.1 | NP_631879.1 | 4 - 5 | 536 | 105 | |
GenBank mRNA | AB079132.1 | - | 4 - 5 | 536 | 105 | |
AK048720.1 | - | 4 - 5 | 578 | 105 | ||
AK135838.1 | - | 4 - 5 | 594 | 105 | ||
AK159335.1 | - | 4 - 5 | 630 | 106 | ||
AK167788.1 | - | 3 - 4 | 213 | 106 | ||
AK171266.1 | - | 3 - 4 | 291 | 106 | ||
BC023432.1 | - | 2 - 3 | 186 | 5029821 | 105 | |
BC072579.1 | - | 4 - 5 | 803 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |