Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290634.1 | NP_001277563.1 | 5 - 6 | 810 | 96 | |
NM_139297.6 | NP_647458.1 | 5 - 6 | 1000 | 96 | ||
GenBank mRNA | AF424698.1 | - | 5 - 6 | 848 | 96 | |
AK033445.1 | - | 5 - 6 | 847 | 96 | ||
AK153885.1 | - | 4 - 5 | 1943 | 96 | ||
AK218540.1 | - | 2 - 3 | 168 | 96 | ||
BC023810.1 | - | 5 - 6 | 851 | 5353993 | 96 | |
BC025585.1 | - | 5 - 6 | 755 | 5324928 | 96 | |
BC026626.1 | - | 4 - 5 | 537 | 96 | ||
BC061208.1 | - | 5 - 6 | 887 | 30250244 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |