Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144500.3 | NP_653083.2 | 4 - 5 | 463 | 84 | |
GenBank mRNA | AK048537.1 | - | 4 - 5 | 383 | 84 | |
AK145292.1 | - | 4 - 5 | 495 | 84 | ||
AK161695.1 | - | 4 - 5 | 402 | 84 | ||
AK166059.1 | - | 4 - 5 | 442 | 84 | ||
AK170306.1 | - | 4 - 5 | 447 | 84 | ||
AK170669.1 | - | 4 - 5 | 464 | 84 | ||
BC026804.1 | - | 4 - 5 | 470 | 4159217 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |